Thermo Scientific Chemicals

2,4,7-Triamino-6-phenylpteridine, 98%, Thermo Scientific Chemicals

Catalog number: B20044.22
100 g, Each
Thermo Scientific Chemicals

2,4,7-Triamino-6-phenylpteridine, 98%, Thermo Scientific Chemicals

Catalog number: B20044.22
100 g, Each
Quantity
Catalog number: B20044.22
also known as B20044-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
396-01-0
IUPAC Name
6-phenylpteridine-2,4,7-triamine
Molecular Formula
C12H11N7
InChI Key
FNYLWPVRPXGIIP-UHFFFAOYSA-N
SMILES
NC1=NC(N)=C2N=C(C(N)=NC2=N1)C1=CC=CC=C1
Form
Powder
Appearance (Color)
Yellow
Assay (HPLC)
≥97.5%
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

Weak diuretic with potassium sparing properties that blocks sodium reuptake in the kidneys

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text