4-Bromo-2-ethyl-1-iodobenzene, 97%, stab. with copper, Thermo Scientific Chemicals
4-Bromo-2-ethyl-1-iodobenzene, 97%, stab. with copper, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-2-ethyl-1-iodobenzene, 97%, stab. with copper, Thermo Scientific Chemicals

Have Questions?
Quantity:
10 g
Catalog number B20052.09
also known as B20052-09
Price (USD)/ Each
185.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
185.00
Add to cart
4-Bromo-2-ethyl-1-iodobenzene, 97%, stab. with copper, Thermo Scientific Chemicals
Catalog numberB20052.09
Price (USD)/ Each
185.00
-
Add to cart
Chemical Identifiers
CAS175278-30-5
IUPAC Name4-bromo-2-ethyl-1-iodobenzene
Molecular FormulaC8H8BrI
InChI KeyJWKQXPHYKRQLEJ-UHFFFAOYSA-N
SMILESCCC1=C(I)C=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to brown
Refractive Index1.6315-1.6365 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only