trans-4-Methylcyclohexanol, 98%, Thermo Scientific Chemicals
trans-4-Methylcyclohexanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-4-Methylcyclohexanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B20054.06
also known as B20054-06
Price (USD)/ Each
95.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
95.10
Add to cart
trans-4-Methylcyclohexanol, 98%, Thermo Scientific Chemicals
Catalog numberB20054.06
Price (USD)/ Each
95.10
-
Add to cart
Chemical Identifiers
CAS7731-29-5
IUPAC Name4-methylcyclohexan-1-ol
Molecular FormulaC7H14O
InChI KeyMQWCXKGKQLNYQG-UHFFFAOYSA-N
SMILESCC1CCC(O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4545-1.4580 @ 20?C
FormLiquid
trans-4-Methylcyclohexanol is used to prepare 4-methylcyclohexyl nitrite to study the laser-induced fluorescence (LIF) excitation spectra of the 4-methylcyclohexoxy and d11-cyclohexoxy radicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Methylcyclohexanol is used to prepare 4-methylcyclohexyl nitrite to study the laser-induced fluorescence (LIF) excitation spectra of the 4-methylcyclohexoxy and d11-cyclohexoxy radicals.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Lei Zhang, et al. Laser-induced fluorescence spectra of 4-methylcyclohexoxy radical and perdeuterated cyclohexoxy radical and direct kinetic studies of their reactions with O2.J. Phys. Chem. A.2005,109(41), 9232-9240.
  2. Miyazawa M, et al. Biological enantioselective reduction of methylcyclohexanones by Glomerella cingulata.J. Chem. Technol. Biotechnol.1996,65(2), 171-175.