Thermo Scientific Chemicals

trans-4-Methylcyclohexanol, 98%, Thermo Scientific Chemicals

Catalog number: B20054.14
25 g, Each
Thermo Scientific Chemicals

trans-4-Methylcyclohexanol, 98%, Thermo Scientific Chemicals

Catalog number: B20054.14
25 g, Each
Quantity
Catalog number: B20054.14
also known as B20054-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7731-29-5
IUPAC Name
4-methylcyclohexan-1-ol
Molecular Formula
C7H14O
InChI Key
MQWCXKGKQLNYQG-UHFFFAOYSA-N
SMILES
CC1CCC(O)CC1
Assay (GC)
>97.5%
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4545-1.4580 @ 20?C
Form
Liquid

Description

trans-4-Methylcyclohexanol is used to prepare 4-methylcyclohexyl nitrite to study the laser-induced fluorescence (LIF) excitation spectra of the 4-methylcyclohexoxy and d11-cyclohexoxy radicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Methylcyclohexanol is used to prepare 4-methylcyclohexyl nitrite to study the laser-induced fluorescence (LIF) excitation spectra of the 4-methylcyclohexoxy and d11-cyclohexoxy radicals.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text