Tetramethylthiuram disulfide, 97%, Thermo Scientific Chemicals
Tetramethylthiuram disulfide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetramethylthiuram disulfide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number B20063.22
also known as B20063-22
Price (USD)/ Each
24.90
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
24.90
Add to cart
Tetramethylthiuram disulfide, 97%, Thermo Scientific Chemicals
Catalog numberB20063.22
Price (USD)/ Each
24.90
-
Add to cart
Chemical Identifiers
CAS137-26-8
IUPAC NameN,N-dimethyl[(dimethylcarbamothioyl)disulfanyl]carbothioamide
Molecular FormulaC6H12N2S4
InChI KeyKUAZQDVKQLNFPE-UHFFFAOYSA-N
SMILESCN(C)C(=S)SSC(=S)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to cream
Assay (HPLC)≥96.0%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only