Thermo Scientific Chemicals

2-Bromo-1-butene, 97%, Thermo Scientific Chemicals

Catalog number: B20067.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-1-butene, 97%, Thermo Scientific Chemicals

Catalog number: B20067.06
5 g, Each
Quantity
Catalog number: B20067.06
also known as B20067-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23074-36-4
IUPAC Name
2-bromobut-1-ene
Molecular Formula
C4H7Br
InChI Key
HQMXRIGBXOFKIU-UHFFFAOYSA-N
SMILES
CCC(Br)=C
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4500-1.4550 @ 20?C
Assay (GC)
≥96.0%

Description

2-Bromo-1-butene is used to produce 2-Ethyl-acrylic acid. It acts as a photolytic precursor for 1-buten-2-yl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-1-butene is used to produce 2-Ethyl-acrylic acid. It acts as a photolytic precursor for 1-buten-2-yl.

Solubility
Soluble in ethanol, acetone and benzene. Not miscible in water.

Notes
Store away from air. Store under dry inert gas. Incompatible with oxidizing agents and air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text