4-Bromo-3-methyl-1H-pyrazole, 97%, Thermo Scientific Chemicals
4-Bromo-3-methyl-1H-pyrazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-3-methyl-1H-pyrazole, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number B20069.06
also known as B20069-06
Price (USD)/ Each
147.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
147.00
Add to cart
4-Bromo-3-methyl-1H-pyrazole, 97%, Thermo Scientific Chemicals
Catalog numberB20069.06
Price (USD)/ Each
147.00
-
Add to cart
Chemical Identifiers
CAS13808-64-5
IUPAC Name4-bromo-5-methyl-1H-pyrazole
Molecular FormulaC4H5BrN2
InChI KeyIXQPRETWBGVNPJ-UHFFFAOYSA-N
SMILESCC1=C(Br)C=NN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Melting Point (clear melt)70.5-79.5?C
FormPowder
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only