Tetrachloroethylene, 99%, Thermo Scientific Chemicals
Tetrachloroethylene, 99%, Thermo Scientific Chemicals
Thermo Scientific™

Tetrachloroethylene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number B20089.AP
also known as B20089-AP
Price (USD)/ Each
51.65
Online exclusive
56.90 
Save 5.25 (9%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
51.65
Online exclusive
56.90 
Save 5.25 (9%)
Add to cart
Tetrachloroethylene, 99%, Thermo Scientific Chemicals
Catalog numberB20089.AP
Price (USD)/ Each
51.65
Online exclusive
56.90 
Save 5.25 (9%)
-
Add to cart
Chemical Identifiers
CAS127-18-4
IUPAC Nametetrachloroethene
Molecular FormulaC2Cl4
InChI KeyCYTYCFOTNPOANT-UHFFFAOYSA-N
SMILESClC(Cl)=C(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Color (APHA)≤15 (U.S. specification)
Assay (GC)≥98.5% (non-U.S. specification)
Assay from Suppliers CofA≥98.5% (U.S.specification)
View more
Tetrachloroethylene is used as a solvent. It is involved in dry cleaning as well as in degreasing of metal parts. It finds application as a neutrino detector. It is utilized as a paint stripper and as a spot remover in consumer products. It acts as a precursor for the manufacture of other chemicals. It plays an important role in water repellants, paint removers, printing inks, glues, sealants, polishes and lubricants. As a film-forming electrolyte additive, it is used in the manufacture of lithium ion batteries and is actively involved as an extraction solvent in the determination of oil and grease in water by FT-IR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrachloroethylene is used as a solvent. It is involved in dry cleaning as well as in degreasing of metal parts. It finds application as a neutrino detector. It is utilized as a paint stripper and as a spot remover in consumer products. It acts as a precursor for the manufacture of other chemicals. It plays an important role in water repellants, paint removers, printing inks, glues, sealants, polishes and lubricants. As a film-forming electrolyte additive, it is used in the manufacture of lithium ion batteries and is actively involved as an extraction solvent in the determination of oil and grease in water by FT-IR.

Solubility
Miscible with alcohol, ether, chloroform, benzene and hexane. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, alkali metals and aluminum.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Qiu, Z.; Yang, W.; He, L.; Zhao, Z.; Lu, S.; Sui, Q. Characteristics and influencing factors of tetrachloroethylene sorption-desorption on soil and its components. Chemosphere 2015, 144, 895-901.
  2. Petrucci, E.; Scarsella, M.; De Filippis, P.; Di Palma, L. Tetrachloroethene recovery and hazard reduction of spent powders from dry cleaning process. Waste Manage. Res. 2015, 33 (4), 339-344.