Thermo Scientific Chemicals

1,3-Propanedisulfonic acid disodium salt, 99% (dry wt.), Thermo Scientific Chemicals

Catalog number: B20092.06
5 g, Each
Thermo Scientific Chemicals

1,3-Propanedisulfonic acid disodium salt, 99% (dry wt.), Thermo Scientific Chemicals

Catalog number: B20092.06
5 g, Each
Quantity
Catalog number: B20092.06
also known as B20092-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
36589-58-9
IUPAC Name
disodium propane-1,3-disulfonate
Molecular Formula
C3H6Na2O6S2
InChI Key
DKGJFKPIUSHDIT-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]S(=O)(=O)CCCS([O-])(=O)=O
Sulfated Ash
56.4-58.1%
Form
Powder
Loss on Drying
<5.0% (200?C/constant wt.)
Appearance (Color)
White
Assay (ex Sulphated Ash)
>98.5 to <101.5% (dry wt. basis)

Description

It is used as a reactant involved in synthesis of cadmium(II) bipyridine complex metal-organic frameworks with alkanedisulfonates, alkylenedisulfonate inorganic-organic hybrid materials and glycosaminoglycan mimetic involved in pharmacological studies for the treatment of amyloid A amyloidosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reactant involved in synthesis of cadmium(II) bipyridine complex metal-organic frameworks with alkanedisulfonates, alkylenedisulfonate inorganic-organic hybrid materials and glycosaminoglycan mimetic involved in pharmacological studies for the treatment of amyloid A amyloidosis.

Solubility
Partially soluble in cold water, hot water.

Notes
Hygroscopic. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text