1,4-Dioxane-2,5-dione, 97%, Thermo Scientific Chemicals
1,4-Dioxane-2,5-dione, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Dioxane-2,5-dione, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B20096.14
also known as B20096-14
Price (USD)/ Each
262.65
Online exclusive
292.00 
Save 29.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
262.65
Online exclusive
292.00 
Save 29.35 (10%)
Add to cart
1,4-Dioxane-2,5-dione, 97%, Thermo Scientific Chemicals
Catalog numberB20096.14
Price (USD)/ Each
262.65
Online exclusive
292.00 
Save 29.35 (10%)
-
Add to cart
Chemical Identifiers
CAS502-97-6
IUPAC Name1,4-dioxane-2,5-dione
Molecular FormulaC4H4O4
InChI KeyRKDVKSZUMVYZHH-UHFFFAOYSA-N
SMILESO=C1COC(=O)CO1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or granules
Identification (FTIR)Conforms
Acid-base back titration≥96.0 to ≤104.0%
Appearance (Color)White
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only