Thermo Scientific Chemicals

4-Bromo-2-fluorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: B20118.14
25 g, Each
Thermo Scientific Chemicals

4-Bromo-2-fluorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: B20118.14
25 g, Each
Quantity
Catalog number: B20118.14
also known as B20118-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112704-79-7
IUPAC Name
4-bromo-2-fluorobenzoate
Molecular Formula
C7H3BrFO2
InChI Key
ZQQSRVPOAHYHEL-UHFFFAOYSA-M
SMILES
[O-]C(=O)C1=CC=C(Br)C=C1F
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Powder
Melting Point (clear melt)
209.0-215.0?C
Appearance (Color)
White to pale cream
Assay (HPLC)
≥97.5%

Description

4-Bromo-2-fluorobenzoic acid may be used in the synthesis of biaryl intermediates by palladium-mediated coupling with various aryl boronic acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-fluorobenzoic acid may be used in the synthesis of biaryl intermediates by palladium-mediated coupling with various aryl boronic acids

Solubility
Solubility in methanol, very faint turbidity.

Notes
Store away from oxidizing agents, reducing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text