Ethyl 2-oxo-4-phenylbutyrate, 90+%, Thermo Scientific Chemicals
Ethyl 2-oxo-4-phenylbutyrate, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-oxo-4-phenylbutyrate, 90+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number B20148.18
also known as B20148-18
Price (USD)/ Each
158.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
158.00
Add to cart
Ethyl 2-oxo-4-phenylbutyrate, 90+%, Thermo Scientific Chemicals
Catalog numberB20148.18
Price (USD)/ Each
158.00
-
Add to cart
Chemical Identifiers
CAS64920-29-2
IUPAC Nameethyl 2-oxo-4-phenylbutanoate
Molecular FormulaC12H14O3
InChI KeySTPXIOGYOLJXMZ-UHFFFAOYSA-N
SMILESCCOC(=O)C(=O)CCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥90.0%
Identification (FTIR)Conforms
Refractive Index1.4635-1.5435 @ 20?C
Appearance (Color)Clear yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only