Thermo Scientific Chemicals

1-Penten-3-ol, 97%, Thermo Scientific Chemicals

Catalog number: B20155.06
5 g, Each
Thermo Scientific Chemicals

1-Penten-3-ol, 97%, Thermo Scientific Chemicals

Catalog number: B20155.06
5 g, Each
Quantity
Catalog number: B20155.06
also known as B20155-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
616-25-1
IUPAC Name
pent-1-en-3-ol
Molecular Formula
C5H10O
InChI Key
VHVMXWZXFBOANQ-UHFFFAOYNA-N
SMILES
CCC(O)C=C
Refractive Index
1.4220-1.4270 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms

Description

1-Penten-3-ol is used as a flavor and a fragrance. It can be used for lift with cucumber, melon, berry, horseradish, strawberry, and tomato.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Penten-3-ol is used as a flavor and a fragrance. It can be used for lift with cucumber, melon, berry, horseradish, strawberry, and tomato.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text