Methyl 2-chloro-4-nitrobenzoate, 98%, Thermo Scientific Chemicals
Methyl 2-chloro-4-nitrobenzoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 2-chloro-4-nitrobenzoate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B20169.06
also known as B20169-06
Price (USD)/ Each
48.20
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
48.20
Add to cart
Methyl 2-chloro-4-nitrobenzoate, 98%, Thermo Scientific Chemicals
Catalog numberB20169.06
Price (USD)/ Each
48.20
-
Add to cart
Chemical Identifiers
CAS13324-11-3
IUPAC Namemethyl 2-chloro-4-nitrobenzoate
Molecular FormulaC8H6ClNO4
InChI KeyPICNSXCJRMYANX-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C=C1Cl)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)73.0-79.0?C
Appearance (Color)White to pale cream
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only