Thermo Scientific Chemicals

N,N-Dimethylethylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: B20182.18
50 g, Each
Thermo Scientific Chemicals

N,N-Dimethylethylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: B20182.18
50 g, Each
Quantity
Catalog number: B20182.18
also known as B20182-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-00-9
IUPAC Name
(2-aminoethyl)dimethylamine
Molecular Formula
C4H12N2
InChI Key
DILRJUIACXKSQE-UHFFFAOYSA-N
SMILES
CN(C)CCN
Assay (GC)
≥96.0%
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4240-1.4290 @ 20?C
Appearance (Color)
Clear colorless to pale yellow

Description

It finds its application as an intermediate in the synthesis of Cefotiam. It is also used as a catalyst in the synthesis of allylic nitro compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as an intermediate in the synthesis of Cefotiam. It is also used as a catalyst in the synthesis of allylic nitro compounds.

Solubility
Fully miscible in water.

Notes
Air sensitive & hygroscopic. Store away from oxidizing agents, air and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from electrostatic charges.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text