It finds its application as an intermediate in the synthesis of Cefotiam. It is also used as a catalyst in the synthesis of allylic nitro compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application as an intermediate in the synthesis of Cefotiam. It is also used as a catalyst in the synthesis of allylic nitro compounds.
Solubility
Fully miscible in water.
Notes
Air sensitive & hygroscopic. Store away from oxidizing agents, air and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from electrostatic charges.
RUO – Research Use Only
General References:
- Rui Tamura, et al. Facile synthesis of allylic nitro compounds by N,N-dimethylethylenediamine-catalyzed condensation of aliphatic and alicyclic ketones with primary nitroalkanes.J. Org. Chem.,1986,51(23), 4368-4375.
- Thomas M. McDonald, et al. Enhanced carbon dioxide capture upon incorporation of N,N'-dimethylethylenediamine in the metal-organic framework CuBTTri.JChem. Sci.,2011,2(10), 2022-2028.