Cyclopropylcarboxamidine hydrochloride, 97%, Thermo Scientific Chemicals
Cyclopropylcarboxamidine hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopropylcarboxamidine hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B20185.14
also known as B20185-14
Price (USD)/ Each
546.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
546.00
Add to cart
Cyclopropylcarboxamidine hydrochloride, 97%, Thermo Scientific Chemicals
Catalog numberB20185.14
Price (USD)/ Each
546.00
-
Add to cart
Chemical Identifiers
CAS57297-29-7
IUPAC Namehydrogen cyclopropanecarboximidamide chloride
Molecular FormulaC4H9ClN2
InChI KeyJRYOZJIRAVZGMV-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC(=N)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)≤1.5%
Melting Point (clear melt)122.0-131.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only