Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)phenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: B20206.03
1 g, Each
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)phenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: B20206.03
1 g, Each
Quantity
Catalog number: B20206.03
also known as B20206-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23165-29-9
IUPAC Name
1-isothiocyanato-3,5-bis(trifluoromethyl)benzene
Molecular Formula
C9H3F6NS
InChI Key
FXOSSGVJGGNASE-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC(=CC(=C1)N=C=S)C(F)(F)F
Identification (FTIR)
Conforms
Form
Liquid
Refractive Index
1.4975-1.5015 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%

Description

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from alcohols, amines and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from water/moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text