Thermo Scientific Chemicals

Ethylidenecyclopentane, 90+%, Thermo Scientific Chemicals

Catalog number: B20209.06
5 g, Each
Thermo Scientific Chemicals

Ethylidenecyclopentane, 90+%, Thermo Scientific Chemicals

Catalog number: B20209.06
5 g, Each
Quantity
Catalog number: B20209.06
also known as B20209-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2146-37-4
IUPAC Name
ethylidenecyclopentane
Molecular Formula
C7H12
InChI Key
VONKRKBGTZDZNV-UHFFFAOYSA-N
SMILES
CC=C1CCCC1
Refractive Index
1.4095-1.4895 @ 20?C
Assay (GC)
≥90.0%
Appearance (Color)
Clear colorless
Form
Liquid

Description

It is used to make other organic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to make other organic chemicals.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text