Allylcyclopentane, 97%, Thermo Scientific Chemicals
Allylcyclopentane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Allylcyclopentane, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number B20228.06
also known as B20228-06
Price (USD)/ Each
65.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
65.90
Add to cart
Allylcyclopentane, 97%, Thermo Scientific Chemicals
Catalog numberB20228.06
Price (USD)/ Each
65.90
-
Add to cart
Chemical Identifiers
CAS3524-75-2
IUPAC Name(prop-2-en-1-yl)cyclopentane
Molecular FormulaC8H14
InChI KeyNHIDGVQVYHCGEK-UHFFFAOYSA-N
SMILESC=CCC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4395-1.4445 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only