Thermo Scientific Chemicals

Methyl N-methylanthranilate, 98%, Thermo Scientific Chemicals

Catalog number: B20233.18
50 g, Each
Thermo Scientific Chemicals

Methyl N-methylanthranilate, 98%, Thermo Scientific Chemicals

Catalog number: B20233.18
50 g, Each
Quantity
Catalog number: B20233.18
also known as B20233-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
85-91-6
IUPAC Name
methyl 2-(methylamino)benzoate
Molecular Formula
C9H11NO2
InChI Key
GVOWHGSUZUUUDR-UHFFFAOYSA-N
SMILES
CNC1=CC=CC=C1C(=O)OC
Appearance (Color)
Clear pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.5780-1.5820 @ 20?C

Description

Methyl N-methylanthranilate is a fragrance ingredient used in decorative cosmetics, fine fragrances, shampoos, toilet soaps and other toiletries as well as in non-cosmetic products such as household cleaners and detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl N-methylanthranilate is a fragrance ingredient used in decorative cosmetics, fine fragrances, shampoos, toilet soaps and other toiletries as well as in non-cosmetic products such as household cleaners and detergents.

Solubility
Miscible in all proportions with ethanol 96%, DMSO and diethyl ether.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text