Thermo Scientific Chemicals

Diethyl phenylmalonate, 98%, Thermo Scientific Chemicals

Catalog number: B20234.30
250 g, Each
Thermo Scientific Chemicals

Diethyl phenylmalonate, 98%, Thermo Scientific Chemicals

Catalog number: B20234.30
250 g, Each
Quantity
Catalog number: B20234.30
also known as B20234-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
83-13-6
IUPAC Name
1,3-diethyl 2-phenylpropanedioate
Molecular Formula
C13H16O4
InChI Key
FGYDHYCFHBSNPE-UHFFFAOYSA-N
SMILES
CCOC(=O)C(C(=O)OCC)C1=CC=CC=C1
Identification (FTIR)
Conforms
Form
Liquid
Refractive Index
1.4885-1.4935 @ 20°C
Appearance (Color)
Clear colorless to yellow to orange
Assay (GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text