1-Acetoxy-2-methoxybenzene, 98%, Thermo Scientific Chemicals
1-Acetoxy-2-methoxybenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Acetoxy-2-methoxybenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number B20242.22
also known as B20242-22
Price (USD)/ Each
96.10
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
96.10
Add to cart
1-Acetoxy-2-methoxybenzene, 98%, Thermo Scientific Chemicals
Catalog numberB20242.22
Price (USD)/ Each
96.10
-
Add to cart
Chemical Identifiers
CAS15212-03-0
IUPAC Name2-methoxyphenyl acetate
Molecular FormulaC9H10O3
InChI KeyBHJHPYFAYGAPLS-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5120-1.5160 @ 20?C
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only