2,2-Dimethyl-3-hexanol, 97%, Thermo Scientific Chemicals
2,2-Dimethyl-3-hexanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2-Dimethyl-3-hexanol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B20244.14
also known as B20244-14
Price (USD)/ Each
288.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
288.00
Add to cart
2,2-Dimethyl-3-hexanol, 97%, Thermo Scientific Chemicals
Catalog numberB20244.14
Price (USD)/ Each
288.00
-
Add to cart
Chemical Identifiers
CAS4209-90-9
IUPAC Name2,2-dimethylhexan-3-ol
Molecular FormulaC8H18O
InChI KeyPFHLGQKVKALLMD-UHFFFAOYNA-N
SMILESCCCC(O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4255-1.4305 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only