Thermo Scientific Chemicals

Methyl 4-bromo-3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: B20249.88
2.5 g, Each
Thermo Scientific Chemicals

Methyl 4-bromo-3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: B20249.88
2.5 g, Each
Quantity
Catalog number: B20249.88
also known as B20249-88
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95201-93-7
IUPAC Name
(2Z)-4-bromo-2-[hydroxy(methoxy)methylidene]-2,3-dihydrothiophen-3-one
Molecular Formula
C6H5BrO3S
InChI Key
YCTROWLDGFWISA-WAYWQWQTSA-N
SMILES
COC(O)=C1/SC=C(Br)C1=O
Assay (GC)
≥96.0%
Appearance (Color)
White to cream or pale orange to pale brown or pink/brown to pink/red
Form
Powder
Melting Point (clear melt)
76-82?C
Identification (FTIR)
Conforms

Description

The synthesis of the thienopyranone scaffold began with the alkylation of methyl 4-bromo-3-hydroxythiophene-2-carboxylate with N-acetylmorpholine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The synthesis of the thienopyranone scaffold began with the alkylation of methyl 4-bromo-3-hydroxythiophene-2-carboxylate with N-acetylmorpholine.

Solubility
Sparingly Soluble in water 0.95 g/L @ 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, bases, reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text