Thermo Scientific Chemicals

Copper(II) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: B20253.22
100 g, Each
Thermo Scientific Chemicals

Copper(II) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: B20253.22
100 g, Each
Quantity
Catalog number: B20253.22
also known as B20253-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
34946-82-2
IUPAC Name
copper(II) triflate
Molecular Formula
C2H2CuF6O4S2
InChI Key
TXWAGEFXPDWAQH-UHFFFAOYSA-N
SMILES
[Cu].FC(F)(F)S(=O)=O.FC(F)(F)S(=O)=O
Appearance (Color)
White to pale cream to pale green or pale blue to pale gray
Assay (Iodometric Titration)
≥97.5 to ≤102.5%
Form
Powder

Description

Copper(II) trifluoromethanesulfonate acts as a catalyst in Diels-Alder reaction and cyclopropanation reactions. It is also used as a reagent for oligosaccharide synthesis. It acts as a Lewis acid. It is also employed in the dimerization of ketone enolates and tetramethylsilane enol ethers. It is used to prepare cis vinylic sulfones and oxazoles from alkynyl sulfones and ketones respectively. Further, it plays an important role for cyclization of dienolates. In addition to this, it is useful for dehydration of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) trifluoromethanesulfonate acts as a catalyst in Diels-Alder reaction and cyclopropanation reactions. It is also used as a reagent for oligosaccharide synthesis. It acts as a Lewis acid. It is also employed in the dimerization of ketone enolates and tetramethylsilane enol ethers. It is used to prepare cis vinylic sulfones and oxazoles from alkynyl sulfones and ketones respectively. Further, it plays an important role for cyclization of dienolates. In addition to this, it is useful for dehydration of alcohols.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text