Thermo Scientific Chemicals

1-Hexene, 98%, Thermo Scientific Chemicals

Catalog number: B20271.AE
100 mL, Each
Thermo Scientific Chemicals

1-Hexene, 98%, Thermo Scientific Chemicals

Catalog number: B20271.AE
100 mL, Each
Quantity
Catalog number: B20271.AE
also known as B20271-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
592-41-6
IUPAC Name
hex-1-ene
Molecular Formula
C6H12
InChI Key
LIKMAJRDDDTEIG-UHFFFAOYSA-N
SMILES
CCCCC=C
Assay (GC)
≥97.5%
Form
Liquid
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Refractive Index
1.3855-1.3900 @ 20?C

Description

1-Hexene is used as a solvent, paint thinner and a medium for conducting chemical reactions. It is a commoner and used in the preparation of both low density and high density polyethylenes. It acts as an adhesives, lubricants, lubricant additives and sealant chemicals. Further, it is used in the preparation of hex-1-en-3-one, heptanal and heptanoic acid. It is also employed in the synthesis of linear plasticizers, oxo-alcohols, motor fuels, automotive additives and biodegradable surfactants. In addition to this, it is used in the preparation of mercaptans, flavors and fragrances, alkyl metals, halides and alkyl silanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexene is used as a solvent, paint thinner and a medium for conducting chemical reactions. It is a commoner and used in the preparation of both low density and high density polyethylenes. It acts as an adhesives, lubricants, lubricant additives and sealant chemicals. Further, it is used in the preparation of hex-1-en-3-one, heptanal and heptanoic acid. It is also employed in the synthesis of linear plasticizers, oxo-alcohols, motor fuels, automotive additives and biodegradable surfactants. In addition to this, it is used in the preparation of mercaptans, flavors and fragrances, alkyl metals, halides and alkyl silanes.

Solubility
Miscible with water, ethanol, ethyl ether, benzene and petroleum ether.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text