Thermo Scientific Chemicals

2',4'-Dihydroxy-3'-methylacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: B20272.14
25 g, Each
Thermo Scientific Chemicals

2',4'-Dihydroxy-3'-methylacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: B20272.14
25 g, Each
Quantity
Catalog number: B20272.14
also known as B20272-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10139-84-1
IUPAC Name
1-(2,4-dihydroxy-3-methylphenyl)ethan-1-one
Molecular Formula
C9H10O3
InChI Key
KMTLZBUHQPQFAV-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=C(O)C(C)=C1O
Form
Powder
Appearance (Color)
Dark cream to pale brown
Assay (GC)
≥97.5%

Description

chemo selective benzylation of 2,4-dihydroxy-3-methylacetophenone gave the 4-benzyloxy derivative. 4- Ethyl-6 methyl resorcinol was prepared by Clemmensen reduction of 4-dihydroxy-3- methylacetophenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
chemo selective benzylation of 2,4-dihydroxy-3-methylacetophenone gave the 4-benzyloxy derivative. 4- Ethyl-6 methyl resorcinol was prepared by Clemmensen reduction of 4-dihydroxy-3- methylacetophenone.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text