Thermo Scientific Chemicals

cis-3-Methylcyclohexanol, 95%, Thermo Scientific Chemicals

Catalog number: B20288.06
5 g, Each
Thermo Scientific Chemicals

cis-3-Methylcyclohexanol, 95%, Thermo Scientific Chemicals

Catalog number: B20288.06
5 g, Each
Quantity
Catalog number: B20288.06
also known as B20288-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24965-90-0
IUPAC Name
3-methylcyclohexan-1-ol
Molecular Formula
C7H14O
InChI Key
HTSABYAWKQAHBT-UHFFFAOYNA-N
SMILES
CC1CCCC(O)C1
Appearance (Color)
Clear colorless
Assay (GC)
≥94.0%
Refractive Index
1.4540-1.4600 @ 20?C
Form
Liquid

Description

cis-3-Methylcyclohexanol is a useful cis methyl cyclohexanol for proteomics research and as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-3-Methylcyclohexanol is a useful cis methyl cyclohexanol for proteomics research and as an intermediate in organic synthesis.

Solubility
Soluble in ethanol, ether. Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text