1,3-Di-tert-butylbenzene is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Di-tert-butylbenzene is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
J T Gerig. Solute-solvent interactions probed by intermolecular NOEs. Journal of Organic Chemistry. 2003, 68 (13), 5244-5248.
James Ray Cozort; James F. OutlawJr., Adrian Hawkins; Samuel Siegel. Dienes as possible intermediates in the catalytic hydrogenation of aromatic hydrocarbons. 2. Dienes derived from 1,3-di-tert-butylbenzene and a rhodium catalyst. J. Org. Chem. 1983, 48 (23), 4190-4197.