5-Bromoindole, 99%, Thermo Scientific Chemicals
5-Bromoindole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromoindole, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B20307.06
also known as B20307-06
Price (USD)/ Each
33.70
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
33.70
Add to cart
5-Bromoindole, 99%, Thermo Scientific Chemicals
Catalog numberB20307.06
Price (USD)/ Each
33.70
-
Add to cart
Chemical Identifiers
CAS10075-50-0
IUPAC Name5-bromo-1H-indole
Molecular FormulaC8H6BrN
InChI KeyVXWVFZFZYXOBTA-UHFFFAOYSA-N
SMILESBrC1=CC=C2NC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
Melting Point (clear melt)87.0-94.0?C
FormPowder
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only