Cyclobutanecarbonyl chloride, 98%, Thermo Scientific Chemicals
Cyclobutanecarbonyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclobutanecarbonyl chloride, 98%, Thermo Scientific Chemicals

 
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B20317.09
also known as B20317-09
Price (USD)/ Each
113.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
113.00
Add to cart
Cyclobutanecarbonyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberB20317.09
Price (USD)/ Each
113.00
-
Add to cart
Chemical Identifiers
CAS5006-22-4
IUPAC Namecyclobutanecarbonyl chloride
Molecular FormulaC5H7ClO
InChI KeyJFWMYCVMQSLLOO-UHFFFAOYSA-N
SMILESClC(=O)C1CCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Refractive Index1.4530-1.4570 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
RUO – Research Use Only