3-Ethylpyridine, 98%, Thermo Scientific Chemicals
3-Ethylpyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Ethylpyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number B20318.14
also known as B20318-14
Price (USD)/ Each
163.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
163.00
Add to cart
3-Ethylpyridine, 98%, Thermo Scientific Chemicals
Catalog numberB20318.14
Price (USD)/ Each
163.00
-
Add to cart
Chemical Identifiers
CAS536-78-7
IUPAC Name3-ethylpyridine
Molecular FormulaC7H9N
InChI KeyMFEIKQPHQINPRI-UHFFFAOYSA-N
SMILESCCC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5000-1.5040 @ 20?C
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only