4-Methyl(thiobenzamide), 97%, Thermo Scientific Chemicals
4-Methyl(thiobenzamide), 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methyl(thiobenzamide), 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number B20320.06
also known as B20320-06
Price (USD)/ Each
196.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
196.00
Add to cart
4-Methyl(thiobenzamide), 97%, Thermo Scientific Chemicals
Catalog numberB20320.06
Price (USD)/ Each
196.00
-
Add to cart
Chemical Identifiers
CAS2362-62-1
IUPAC Name4-methylbenzene-1-carbothioamide
Molecular FormulaC8H9NS
InChI KeyQXYZSNGZMDVLKN-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow to pale brown
Assay (Redox Titration)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
FormPowder
Melting Point (clear melt)163.0-173.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only