Thermo Scientific Chemicals

Palmitic acid, 95%, Thermo Scientific Chemicals

Catalog number: B20322.36
500 g, Each
Thermo Scientific Chemicals

Palmitic acid, 95%, Thermo Scientific Chemicals

Catalog number: B20322.36
500 g, Each
Quantity
Catalog number: B20322.36
also known as B20322-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57-10-3
IUPAC Name
hexadecanoic acid
Molecular Formula
C16H32O2
InChI Key
IPCSVZSSVZVIGE-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCC(O)=O
Appearance (Color)
White to pale cream
Form
Powder or granules or flakes or prills
Assay (Silylated GC)
≥94.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
57.0-70.0°C

Description

A saturated fatty acid. Palmitic acid is a saturated fatty acid used in hair care, cosmetics, soaps, paint, rubber, food, pharmaceuticals, animal feed and textiles. It is used to prepare sodium palmitate which is a natural additive in organic products. It is involved in the preparation of cetyl alcohol utilized in the preparation of detergents and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A saturated fatty acid. Palmitic acid is a saturated fatty acid used in hair care, cosmetics, soaps, paint, rubber, food, pharmaceuticals, animal feed and textiles. It is used to prepare sodium palmitate which is a natural additive in organic products. It is involved in the preparation of cetyl alcohol utilized in the preparation of detergents and cosmetics.

Solubility
Soluble in hot alcohol, acetone, benzene, ethyl ether, amyl acetate, propyl alcohol and chloroform. Slightly soluble in cold alcohol and petroleum ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong reducing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text