1-tert-Butyl-3-ethylbenzene, 98%, Thermo Scientific Chemicals
1-tert-Butyl-3-ethylbenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-tert-Butyl-3-ethylbenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B20325.14
also known as B20325-14
Price (USD)/ Each
277.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
277.00
Add to cart
1-tert-Butyl-3-ethylbenzene, 98%, Thermo Scientific Chemicals
Catalog numberB20325.14
Price (USD)/ Each
277.00
-
Add to cart
Chemical Identifiers
CAS14411-56-4
IUPAC Name1-tert-butyl-3-ethylbenzene
Molecular FormulaC12H18
InChI KeyMUJPTTGNHRHIPH-UHFFFAOYSA-N
SMILESCCC1=CC=CC(=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4910-1.4950 @ 20?C
FormLiquid
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only