Thermo Scientific Chemicals

4-Hydrazinobenzoic acid hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: B20326.09
10 g, Each
Thermo Scientific Chemicals

4-Hydrazinobenzoic acid hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: B20326.09
10 g, Each
Quantity
Catalog number: B20326.09
also known as B20326-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24589-77-3
IUPAC Name
hydrogen 4-hydrazinylbenzoic acid chloride
Molecular Formula
C7H9ClN2O2
InChI Key
XHLQMKQBCHYRLC-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NNC1=CC=C(C=C1)C(O)=O
Appearance (Color)
White to pale cream to pale yellow to pale brown
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

4-Hydrazinobenzoic acid hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It may be used for the preparation of 4-(3-(2-(tert-butoxycarbonylamino)ethyl)-5-hydroxy-1H-pyrazol-1-yl)benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydrazinobenzoic acid hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It may be used for the preparation of 4-(3-(2-(tert-butoxycarbonylamino)ethyl)-5-hydroxy-1H-pyrazol-1-yl)benzoic acid.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text