Formamidine disulfide dihydrochloride is used as an intermediate in agriculture and dyestuff intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Formamidine disulfide dihydrochloride is used as an intermediate in agriculture and dyestuff intermediate.
Solubility
Soluble in water.
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
RUO – Research Use Only
J Li, J.D Miller. Reaction kinetics for gold dissolution in acid thiourea solution using formamidine disulfide as oxidant. Hydrometallurgy. 2002, 63 (3), 215-223.
A.E. Bolzán; I.B. Wakenge; R.C. Salvarezza, A.J. Arvia. Electrochemical response of thiourea and formamidine disulphide on polycrystalline platinum in aqueous 0.5 M sulphuric acid. Journal of Electroanalytical Chemistry. 1999, 475 (2), 181-189.