Oxamic acid could also inhibit lactic dehydrogenase in the intact Ehrlich ascites tumor cell.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Oxamic acid could also inhibit lactic dehydrogenase in the intact Ehrlich ascites tumor cell.
Solubility
Soluble in water 108 mg/mL.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
- Sergi Garcia-Segura; Enric Brillas, Sergi Garcia-Segura; Enric Brillas. Mineralization of the recalcitrant oxalic and oxamic acids by electrochemical advanced oxidation processes using a boron-doped diamond anode. Water Research. 2011, 45 (9), 2975-2984.
- John Papaconstantinou And Sidney P. Colowick. Effects Of Oxamic Acid On The Metabolism Of Ehrlich Ascites Tumor Cells In Vitro. The Journal of Biological Chemistry. 1961, 236, 278-284.