Thermo Scientific Chemicals

Oxamic acid, 98%, Thermo Scientific Chemicals

Catalog number: B20344.22
100 g, Each
Thermo Scientific Chemicals

Oxamic acid, 98%, Thermo Scientific Chemicals

Catalog number: B20344.22
100 g, Each
Quantity
Catalog number: B20344.22
also known as B20344-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
471-47-6
IUPAC Name
carbamoylformic acid
Molecular Formula
C2H3NO3
InChI Key
SOWBFZRMHSNYGE-UHFFFAOYSA-N
SMILES
NC(=O)C(O)=O
Assay (special)
≥97.5 to ≤102.5% (based on elemetal H, C, N)
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Powder

Description

Oxamic acid could also inhibit lactic dehydrogenase in the intact Ehrlich ascites tumor cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxamic acid could also inhibit lactic dehydrogenase in the intact Ehrlich ascites tumor cell.

Solubility
Soluble in water 108 mg/mL.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text