Phosphoenolpyruvic acid monopotassium salt, 99%, Thermo Scientific Chemicals
Phosphoenolpyruvic acid monopotassium salt, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phosphoenolpyruvic acid monopotassium salt, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B20358.06
also known as B20358-06
Price (USD)/ Each
185.65
Online exclusive
206.00 
Save 20.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
185.65
Online exclusive
206.00 
Save 20.35 (10%)
Add to cart
Phosphoenolpyruvic acid monopotassium salt, 99%, Thermo Scientific Chemicals
Catalog numberB20358.06
Price (USD)/ Each
185.65
Online exclusive
206.00 
Save 20.35 (10%)
-
Add to cart
Chemical Identifiers
CAS4265-07-0
IUPAC Namepotassium 2-(hydrogen phosphonatooxy)prop-2-enoic acid
Molecular FormulaC3H4KO6P
InChI KeySOSDSEAIODNVPX-UHFFFAOYSA-M
SMILES[K+].OC(=O)C(=C)OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5% (dry wt. basis)
Water Content (Karl Fischer Titration)≤1.50%
Identification (FTIR)Conforms
Formpowder
Phosphoenolpyruvic acid monopotassium salt is used in glycolysis and gluconeogeneis. It is used for regeneration of ATP from ADP in enzyme-catalyzed syntheses. It is metabolized by an enzyme namely pyruvate kinase to yield pyruvate. It acts as protection against cell injury induced by oxidative stress and against dysfunction in carbohydrate metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphoenolpyruvic acid monopotassium salt is used in glycolysis and gluconeogeneis. It is used for regeneration of ATP from ADP in enzyme-catalyzed syntheses. It is metabolized by an enzyme namely pyruvate kinase to yield pyruvate. It acts as protection against cell injury induced by oxidative stress and against dysfunction in carbohydrate metabolism.

Solubility
Soluble in water. Slightly soluble in methanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Used for regeneration of ATP from ADP in enzyme-catalyzed syntheses: J. Org. Chem., 47, 3765 (1982); 50, 1076 (1985); J. Am. Chem. Soc., 107, 7019 (1985).
  2. Yarman, A.; Schulz, C.; Sygmund, C.; Ludwig, R.; Gorton, L.; Wollenberger, U.; Scheller, F. W. Third Generation ATP Sensor with Enzymatic Analyte Recycling. Electroanalysis 2014, 26 (9), 2043-2048.
  3. Gauss, D.; Schoenenberger, B.; Wohlgemuth, R. Chemical and enzymatic methodologies for the synthesis of enantiomerically pure glyceraldehyde 3-phosphates. Carbohydr. Res. 2014, 389, 18-24.