Thermo Scientific Chemicals

Phosphoenolpyruvic acid monopotassium salt, 99%, Thermo Scientific Chemicals

Catalog number: B20358.14
25 g, Each
Thermo Scientific Chemicals

Phosphoenolpyruvic acid monopotassium salt, 99%, Thermo Scientific Chemicals

Catalog number: B20358.14
25 g, Each
Quantity
Catalog number: B20358.14
also known as B20358-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4265-07-0
IUPAC Name
potassium 2-(hydrogen phosphonatooxy)prop-2-enoic acid
Molecular Formula
C3H4KO6P
InChI Key
SOSDSEAIODNVPX-UHFFFAOYSA-M
SMILES
[K+].OC(=O)C(=C)OP(O)([O-])=O
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.5% (dry wt. basis)
Water Content (Karl Fischer Titration)
≤1.50%
Identification (FTIR)
Conforms
Form
powder

Description

Phosphoenolpyruvic acid monopotassium salt is used in glycolysis and gluconeogeneis. It is used for regeneration of ATP from ADP in enzyme-catalyzed syntheses. It is metabolized by an enzyme namely pyruvate kinase to yield pyruvate. It acts as protection against cell injury induced by oxidative stress and against dysfunction in carbohydrate metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphoenolpyruvic acid monopotassium salt is used in glycolysis and gluconeogeneis. It is used for regeneration of ATP from ADP in enzyme-catalyzed syntheses. It is metabolized by an enzyme namely pyruvate kinase to yield pyruvate. It acts as protection against cell injury induced by oxidative stress and against dysfunction in carbohydrate metabolism.

Solubility
Soluble in water. Slightly soluble in methanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text