Thermo Scientific Chemicals

Ethyl 2-chloronicotinate, 98%, Thermo Scientific Chemicals

Catalog number: B20359.09
10 g, Each
Thermo Scientific Chemicals

Ethyl 2-chloronicotinate, 98%, Thermo Scientific Chemicals

Catalog number: B20359.09
10 g, Each
Quantity
Catalog number: B20359.09
also known as B20359-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1452-94-4
IUPAC Name
ethyl 2-chloropyridine-3-carboxylate
Molecular Formula
C8H8ClNO2
InChI Key
PMIMPBYTPPRBGD-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=CN=C1Cl
Form
Liquid
Refractive Index
1.5210-1.5250 @ 20°C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%

Description

Ethyl 2-chloronicotinate,is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-chloronicotinate,is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text