Thermo Scientific Chemicals

2-Amino-2',5-dichlorobenzophenone, 98%, Thermo Scientific Chemicals

Catalog number: B20360.18
50 g, Each
Thermo Scientific Chemicals

2-Amino-2',5-dichlorobenzophenone, 98%, Thermo Scientific Chemicals

Catalog number: B20360.18
50 g, Each
Quantity
Catalog number: B20360.18
also known as B20360-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2958-36-3
IUPAC Name
4-chloro-2-(2-chlorobenzoyl)aniline
Molecular Formula
C13H9Cl2NO
InChI Key
KWZYIAJRFJVQDO-UHFFFAOYSA-N
SMILES
NC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1Cl
Appearance (Color)
Yellow
Assay (GC)
≥97.5%
Form
Powder
Melting Point (clear melt)
86.0-92.0?C

Description

2-Amino-2',5-dichlorobenzophenone is used to produce lorazepam a drug of the benzodiazepine group, used especially to treat anxiety.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-2′,5-dichlorobenzophenone is used to produce lorazepam a drug of the benzodiazepine group, used especially to treat anxiety.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text