Thermo Scientific Chemicals

1-Cyclohexylethanol, 98%, Thermo Scientific Chemicals

Catalog number: B20363.14
25 g, Each
Thermo Scientific Chemicals

1-Cyclohexylethanol, 98%, Thermo Scientific Chemicals

Catalog number: B20363.14
25 g, Each
Quantity
Catalog number: B20363.14
also known as B20363-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1193-81-3
IUPAC Name
1-cyclohexylethan-1-ol
Molecular Formula
C8H16O
InChI Key
JMSUNAQVHOHLMX-UHFFFAOYNA-N
SMILES
CC(O)C1CCCCC1
Refractive Index
1.4640-1.4680 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Form
Liquid or viscous liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text