It is used as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an active pharmaceutical intermediate.
Solubility
Slighlty soluble in aqeous solvents.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Howard Alper.; Khaled Hachem. Phase transfer and rhodium (I) catalyzed isomerization of allylaromatics and cognate systems.Transition Met. Chem.1981,6(4), 219-220.
William A. Bonner.; Frank D. Mango.Kolbe. Electrolyses of 3-Phenyl- and 3,3-Diphenylpropanoic Acids.J. Org. Chem.,1964,29(2), 430-435.