Thermo Scientific Chemicals

2,3,4-Trihydroxybenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: B20371.14
25 g, Each
Thermo Scientific Chemicals

2,3,4-Trihydroxybenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: B20371.14
25 g, Each
Quantity
Catalog number: B20371.14
also known as B20371-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
610-02-6
IUPAC Name
2,3,4-trihydroxybenzoic acid
Molecular Formula
C7H6O5
InChI Key
BRRSNXCXLSVPFC-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(O)C(O)=C1O
Form
Powder
Appearance (Color)
Pale cream to pale gray to gray
Assay (Silylated GC)
≥96.0%
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

2,3,4-Trihydroxybenzoic acid is used as a reagent during catalytic spectrophotometric determination of vanadium in natural and sea waters and used in the pharmaceuticals. It is a internal standard in separation of phenolic acids by HPLC with photodiode array detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4-Trihydroxybenzoic acid is used as a reagent during catalytic spectrophotometric determination of vanadium in natural and sea waters and used in the pharmaceuticals. It is a internal standard in separation of phenolic acids by HPLC with photodiode array detection.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text