Thermo Scientific Chemicals

2,3,6-Trifluorophenol, 98%, Thermo Scientific Chemicals

Catalog number: B20379.03
1 g, Each
Thermo Scientific Chemicals

2,3,6-Trifluorophenol, 98%, Thermo Scientific Chemicals

Catalog number: B20379.03
1 g, Each
Quantity
Catalog number: B20379.03
also known as B20379-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
113798-74-6
Appearance (Color)
White to cream
Form
Fused solid or lumpy powder
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
28.0-36.0°C

Description

2,3,6-Trifluorophenol may be employed as model compound to investigate the site-selective functionalization of halogen-bearing phenols by organometallic approach.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,6-Trifluorophenol may be employed as model compound to investigate the site-selective functionalization of halogen-bearing phenols by organometallic approach.

Solubility
Soluble in water (47.267 mg/L est.)

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text