Thermo Scientific Chemicals

4-Dimethylaminobenzophenone, 98%, Thermo Scientific Chemicals

Catalog number: B20389.06
5 g, Each
Thermo Scientific Chemicals

4-Dimethylaminobenzophenone, 98%, Thermo Scientific Chemicals

Catalog number: B20389.06
5 g, Each
Quantity
Catalog number: B20389.06
also known as B20389-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
530-44-9
IUPAC Name
4-benzoyl-N,N-dimethylaniline
Molecular Formula
C15H15NO
InChI Key
BEUGBYXJXMVRFO-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Form
Powder
Melting Point (clear melt)
87.5-93.5?C
Appearance (Color)
Yellow to green
Assay (GC)
≥97.5%

Description

4-(Dimethylamino)benzophenone is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Dimethylamino)benzophenone is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text