Thermo Scientific Chemicals

Cyclohexyl ethyl ketone, 99%, Thermo Scientific Chemicals

Catalog number: B20409.06
5 g, Each
Thermo Scientific Chemicals

Cyclohexyl ethyl ketone, 99%, Thermo Scientific Chemicals

Catalog number: B20409.06
5 g, Each
Quantity
Catalog number: B20409.06
also known as B20409-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1123-86-0
IUPAC Name
1-cyclohexylpropan-1-one
Molecular Formula
C9H16O
InChI Key
AMHOPTNGSNYSBL-UHFFFAOYSA-N
SMILES
CCC(=O)C1CCCCC1
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Refractive Index
1.4505-1.4545 @ 20?C
Form
Liquid

Description

Cyclohexyl ethyl ketone is used as a intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexyl ethyl ketone is used as a intermediate in organic synthesis.

Solubility
Soluble in organic solvents.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text